missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP4 Protein
A cDNA sequence encoding the FABP4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11836-EC-10ug
Additional Details : Weight : 0.01000kg
Specifications
2167 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
CDAFVGTWKL VSSENFDDYM KEVGVGFATR KVAGMAKPNM IISVNGDVIT IKSESTFKNT EISFILGQEF DEVTADDRKV KSTITLDGGV LVHVQKWDGK STTIKRKRED DKLVVECVMK GVTSTRVYER A | |
Greater than 85.0% as determined by SDS-PAGE. |
FABP4 Protein | |
Research Use Only | |
FABP4 | |
Human | |
His | |
FABP4 protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 6.5. |