missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FAM150A Protein

A DNA sequence encoding the Homo sapiens (Human) FAM150A, was expressed in the hosts and tags indicated.

€ 245.00 - € 3405.00

Specifications

Accession Number NP_997296.1
Gene ID (Entrez) 389658
Name FAM150A Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16030681
View Documents Promotion Details
enQuireBio™
QP7769-EC-100UG
100 μg
€ 770.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16050681
View Documents Promotion Details
enQuireBio™
QP7769-EC-1MG
1 mg
€ 3405.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16040681
View Documents Promotion Details
enQuireBio™
QP7769-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16060681
View Documents Promotion Details
enQuireBio™
QP7769-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16070681
View Documents Promotion Details
enQuireBio™
QP7769-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16080681
View Documents Promotion Details
enQuireBio™
QP7769-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_997296.1
FAM150A Protein
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
389658
Research Use Only
FAM150A
Recombinant Protein
E. coli
RPRGRRGARV TDKEPKPLLF LPAAGAGRTP SGSRSAEIFP RDSNLKDKFI KHFTGPVTFS PECSKHFHRL YYNTRECSTP AYYKRCARLL TRLAVSPLCS QT
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications