Learn More
enQuireBio™ Recombinant Human Fanconi anemia group C Protein
Product Code. 15969399
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 15969399
Brand: enQuireBio™ QP6019ec1mg
A DNA sequence encoding the Homo sapiens (Human) Fanconi anemia group C protein, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_000127.2
Gene ID (Entrez) 2176
Name Fanconi anemia group C Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol FANCC
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MAQDSVDLSC DYQFWMQKLS VWDQASTLET QQDTCLHVAQ FQEFLRKMYE ALKEMDSNTV IERFPTIGQL LAKACWNPFI LAYDESQKIL IWCLCCLINK EPQNSGQSKL NSWIQGVLSH ILSALRFDKE VALFTQGLGY APIDYYPGLL KNMVLSLASE LRENHLNGFN TQRRMAPERV ASLSRVCVPL ITLTDVDPLV EALLICHGRE PQEILQPEFF EAVNEAILLK KISLPMSAVV CLWLRHLPSL EKAMLHLFEK LISSERNCLR RIECFIKDSS LPQAACHPAI FRVVDEMFRC ALLETDGALE IIATIQVFTQ CFVEALEKAS KQLRFALKTY FPYTSPSLAM VLLQDPQDIP RGHWLQTLKH ISELLREAVE DQTHGSCGGP FESWFLFIHF GGWAEMVAEQ LLMSAAEPPT ALLWLLAFYY GPRDGRQQRA QTMVQVKAVL GHLLAMSRSS SLSAQDLQTV AGQGTDTDLR APAQQLIRHL LLNFLLWAPG GHTIAWDVIT LMAHTAEITH EIIGFLDQTL YRWNRLGIES PRSEKLAREL LKELRTQV
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.