missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fc receptor-like protein 1

A DNA sequence encoding the Homo sapiens (Human) Fc receptor-like protein 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7695-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030251

Quantity Price
1 € 245.00 / 10µg
6 € 228.51 / 10µg
Estimated Shipment
04-11-2020
Add to basket


Specifications

Specifications

NP_001004310.2
Fc receptor-like protein 1
10 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
343413
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FCRL6
Recombinant Protein
Yeast
LYLQAWPNPV FEGDALTLRC QGWKNTPLSQ VKFYRDGKFL HFSKENQTLS MGAATVQSRG QYSCSGQVMY IPQTFTQTSE TAMVQVQELF PPPVLSAIPS PEPREGSLVT LRCQTKLHPL RSALRLLFSF HKDGHTLQDR GPHPELCIPG AKEGDSGLYW CEVAPEGGQV QKQSPQLEVR VQAPVSRPVL TLHHGPADPA VGDMVQLLCE AQRGSPPILY SFYLDEKIVG NHSAPCGGTT SLLFPVKSEQ DAGNYSCEAE NSVSRERSEP KKLSLKGSQV LFTPASNW
Videos
SDS
Documents