missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fc receptor-like protein 1

A DNA sequence encoding the Homo sapiens (Human) Fc receptor-like protein 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7876-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031331

Quantity Price
1 € 2645.00 / 1mg
3 € 2469.90 / 1mg
Estimated Shipment
22-10-2020
Add to basket


Specifications

Specifications

NP_112572.1
Fc receptor-like protein 1
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
83417
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FCRL4
Recombinant Protein
Yeast
AHKPVISVHP PWTTFFKGER VTLTCNGFQF YATEKTTWYH RHYWGEKLTL TPGNTLEVRE SGLYRCQARG SPRSNPVRLL FSSDSLILQA PYSVFEGDTL VLRCHRRRKE KLTAVKYTWN GNILSISNKS WDLLIPQASS NNNGNYRCIG YGDENDVFRS NFKIIKIQEL FPHPELKATD SQPTEGNSVN LSCETQLPPE RSDTPLHFNF FRDGEVILSD WSTYPELQLP TVWRENSGSY WCGAETVRGN IHKHSPSLQI HVQRIPVSGV LLETQPSGGQ AVEGEMLVLV CSVAEGTGDT TFSWHREDMQ ESLGRKTQRS LRAELELPAI RQSHAGGYYC TADNSYGPVQ SMVLNVTVRE TPGNRDGL
Videos
Documents