missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FGF basic/FGF2 Protein, CF
Click to view available options
:
500 μg
Description
Measured by its ability to inhibit human FGF-basic-dependent proliferation of NR6R-3T3 mouse fibroblast cells. Rizzino, A. et al. (1988) Cancer Res. 48:4266; Thomas, K. et al. (1987) Methods Enzymol. 147:120; Watson, J.B. et al. (1994) J. Clin. Invest. 94:261. The ED50 for this effect is 0.100-1.00 ng/mL.
Specifications
Specifications
| Accession Number | NP_001997.5 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in HEPES and Sodium Sulfate with Trehalose. |
| Gene ID (Entrez) | 2247 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 18 kDa, under reducing conditions., MolecularWeight-theroretical: 16 kDa |
| Quantity | 500 μg |
| Source | E. coli-derived human FGF basic/FGF2/bFGF protein Ala135-Ser288 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.01 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction