missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FIBP Protein
A DNA sequence encoding the Homo sapiens (Human) FIBP, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_004205.2 |
---|---|
Gene ID (Entrez) | 9158 |
Name | FIBP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15919609
|
enQuireBio™
QP6048-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939609
|
enQuireBio™
QP6048-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929609
|
enQuireBio™
QP6048-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15949609
|
enQuireBio™
QP6048-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15959609
|
enQuireBio™
QP6048-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969609
|
enQuireBio™
QP6048-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004205.2 | |
FIBP Protein | |
Not determined. | |
Full Length of Isoform Short | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9158 | |
Research Use Only | |
FIBP | |
Recombinant Protein | |
E. coli | |
TSELDIFVGN TTLIDEDVYR LWLDGYSVTD AVALRVRSGI LEQTGATAAV LQSDTMDHY RTFHMLERLL HAPPKLLHQL IFQIPPSRQA LLIERYYAFD EAFVREVLGK KLSKGTKKDL DDISTKTGI TLKSCRRQFD NFKRVFKVVE EMRGSLVDNI QQHFLLSDRL ARDYAAIVFF ANNRFETGK KKLQYLSFGD FAFCAELMIQ NWTLGAVDSQ MDDMDMDLDK EFLQDLKELK VLVADKDLL DLHKSLVCTA LRGKLGVFSE MEANFKNLSR GLVNVAAKLT HNKDVRDLFV DLVEKFVEP CRSDHWPLSD VRFFLNQYSA SVHSLDGFRH QALWDRYMGT LRGCLLRLYH D | |
Greater than 90% as determined by SDS-PAGE. |