missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ficolin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ficolin-2, was expressed in the hosts and tags indicated.
€ 202.00 - € 2155.00
Specifications
Accession Number | NP_004099.2 |
---|---|
Gene ID (Entrez) | 2220 |
Name | Ficolin-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088671
|
enQuireBio™
QP9759-YE-100UG |
100 μg |
€ 645.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028681
|
enQuireBio™
QP9759-YE-500UG |
500 μg |
€ 1740.00
500µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098671
|
enQuireBio™
QP9759-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018681
|
enQuireBio™
QP9759-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038681
|
enQuireBio™
QP9759-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008681
|
enQuireBio™
QP9759-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004099.2 | |
Ficolin-2 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2220 | |
Research Use Only | |
FCN2 | |
Recombinant Protein | |
Yeast | |
LQAADTCPEV KMVGLEGSDK LTILRGCPGL PGAPGPKGEA GTNGKRGERG PPGPPGKAGP PGPNGAPGEP QPCLTGPRTC KDLLDRGHFL SGWHTIYLPD CRPLTVLCDM DTDGGGWTVF QRRVDGSVDF YRDWATYKQG FGSRLGEFWL GNDNIHALTA QGTSELRVDL VDFEDNYQFA KYRSFKVADE AEKYNLVLGA FVEGSAGDSL TFHNNQSFST KDQDNDLNTG NCAVMFQGAW WYKNCHVSNL NGRYLRGTHG SFANGINWKS GKGYNYSYKV SEMKVRPA | |
Greater than 90% as determined by SDS-PAGE. |