missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FKBP1B Protein

A cDNA sequence encoding the FKBP1B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10607-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15924398

  • € 4090.00 / 1mg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

2281
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 300 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin.
Human
His
The FKBP1B solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT.
FKBP1B Protein
Research Use Only
FKBP1B
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH RSMGVEIETI SPGDGRTFPK KGQTCVVHYT GMLQNGKKFD SSRDRNKPFK FRIGKQEVIK GFEEGAAQMS LGQRAKLTCT PDVAYGATGH PGVIPPNATL IFDVELLNLE
Greater than 90.0% as determined by SDS-PAGE analysis.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers