missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FLT1 D5 Protein
A cDNA sequence encoding the FLT1 D5 was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 1500.00
Specifications
Gene ID (Entrez) | 2321 |
---|---|
Name | FLT1 D5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | FLT1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954418
|
enQuireBio™
QP10614-2UG |
2 μg |
€ 137.00
2µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944418
|
enQuireBio™
QP10614-10UG |
10 μg |
€ 212.00
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934418
|
enQuireBio™
QP10614-100UG |
100 μg |
€ 1500.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
2321 | |
Research Use Only | |
FLT1 | |
Recombinant Protein | |
Baculovirus | |
MVSYWDTGVL LCALLSCLLL TGSSSGSKLK DPELSLKGTQ HIMQAGQTLH LQCRGEAAHK WSLPEMVSKE SERLSITKSA CGRNGKQFCS TLTLNTAQAN HTGFYSCKYL AVPTSKKKET ESAIYIFISD TGRPFVEMYS EIPEIIHMTE GRELVIPCRV TSPNITVTLK KFPLDTLIPD GKRIIWDSRK GFIISNATYK EIGLLTCEAT VNGHLYKTNY LTHRQTNTII DVQISTPRPV KLLRGHTLVL NCTATTPLNT RVQMTWSYPD EKNKRASVRR RIDQSNSHAN IFYSVLTIDK MQNKDKGLYT CRVRSGPSFK SVNTSVHIYD KAFITVKHRK QQVLETVAGK RSYRLSMKVK AFPSPEVVWL KDGLPATEKS ARYLTRGYSL IIKDVTEEDA GNYTILLSIK QSNVFKNLTA TLIVNVKPQI YEKAVSSFPD PALYPLGSRQ ILTCTAYGIP QPTIKWFWHP CNHNHSEARC DFCSNNEESF ILDADSNMGN RIESITQRMA IIEGKNKMAS TLVVADSRIS GIYICIASNK VGTVGRNISF YITDVPNGFH VNLEKMPTEG EDLKLSCTVN KFLYRDVTWI LLRTVNNRTM HYSISKQKMA ITKEHSITLN LTIMNVSLQD SGTYACRARN VYTGEEILQK KEITIRDQEA PYLLRNLSDH TVAISSSTTL DCHANGVPEP QITWFKNNHK IQQEPGIILG PGSSTLFIER VTEEDEGVYH CKATNQKGSV ESSAYLTVQG TSDKSNLELI TLTCTCVAAT LFWLLLTLFI RKMKRSSSEI KTDYLSIIMD PDEVPLDEQC ERLPYDASKW EFARERLKLG KSLGRGAFGK VVQASAFGIK KSPTCRTVAV KMLKEGATAS EYKALMTELK ILTHIGHHLN VVNLLGACTK QGGPLMVIVE | |
Greater than 90.0% as determined by(a)Analysis by RP-HPLC. (b)Analysis by SDS-PAGE. |
FLT1 D5 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The activity of FLT1 D5 was determined by its ability to abolish the binding of iodinated VEGF to solid surfaces or cell surfaces. The ED50 for this effect is typically 10 ng/ml, corresponding to a specific activity of 100,000IU/mg. In a 13 day CAM-assay sVEGFR-1 is able to inhibit VEGF stimulated sprouting of capillaries at 30 pM. | |
Human | |
Untagged | |
FLT1 D1-5 was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives. |