missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Four and a half LIM domains protein 1

A DNA sequence encoding the Homo sapiens (Human) Four and a half LIM domains protein 3, was expressed in the hosts and tags indicated.

€ 160.00 - € 2385.00

Specifications

Accession Number NP_001230807.1
Gene ID (Entrez) 2275
Name Four and a half LIM domains protein 1
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16089380
View Documents Promotion Details
enQuireBio™
QP7560-EC-100UG
100 μg
€ 530.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16009390
View Documents Promotion Details
enQuireBio™
QP7560-EC-1MG
1 mg
€ 2385.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16099380
View Documents Promotion Details
enQuireBio™
QP7560-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16019390
View Documents Promotion Details
enQuireBio™
QP7560-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16029390
View Documents Promotion Details
enQuireBio™
QP7560-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16039390
View Documents Promotion Details
enQuireBio™
QP7560-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001230807.1
Four and a half LIM domains protein 1
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
2275
Research Use Only
FHL3
Recombinant Protein
E. coli
SESFDCAKCN ESLYGRKYIQ TDSGPYCVPC YDNTFANTCA ECQQLIGHDS RELFYEDRHF HEGCFRCCRC QRSLADEPFT CQDSELLCND CYCSAFSSQC SACGETVMPG SRKLEYGGQT WHEHCFLCSG CEQPLGSRSF VPDKGAHYCV PCYENKFAPR CARCSKTLTQ GGVTYRDQPW HRECLVCTGC QTPLAGQQFT SRDEDPYCVA CFGELFAPKC SSCKRPIVGL GGGKYVSFED RHWHHNCFSC ARCSTSLVGQ GFVPDGDQVL CQGCSQAGP
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications