Learn More
R&D Systems™ Recombinant Human Galectin-3BP/MAC-2BP Protein
Product Code. 18789286
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18789286 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18789286 Supplier R&D Systems™ Supplier No. 2226GAB050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
The Recombinant Human Galectin-3BP/MAC-2BP Protein is derived from NS0. The Recombinant Human Galectin-3BP/MAC-2BP Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q08380.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS and DTT with Trehalose.
Gene ID (Entrez) 3959
Molecular Weight (g/mol) 65 kDa
Name 90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, Galectin3BP, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, LGALS3BP, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Galectin-3 BP/MAC-2 BP protein Val19-Asp585, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.