missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Gamma-crystallin D Protein

A DNA sequence encoding the Homo sapiens (Human) Gamma-crystallin D, was expressed in the hosts and tags indicated.

€ 160.00 - € 2385.00

Specifications

Accession Number NP_008822.2
Gene ID (Entrez) 1421
Name Gamma-crystallin D Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15918349
View Documents Promotion Details
enQuireBio™
QP5871-EC-100UG
100 μg
€ 530.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15938349
View Documents Promotion Details
enQuireBio™
QP5871-EC-1MG
1 mg
€ 2385.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15928349
View Documents Promotion Details
enQuireBio™
QP5871-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15948349
View Documents Promotion Details
enQuireBio™
QP5871-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15958349
View Documents Promotion Details
enQuireBio™
QP5871-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15968349
View Documents Promotion Details
enQuireBio™
QP5871-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_008822.2
Gamma-crystallin D Protein
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
1421
Research Use Only
CRYGD
Recombinant Protein
E. coli
GKITLYEDRG FQGRHYECSS DHPNLQPYLS RCNSARVDSG CWMLYEQPNY SGLQYFLRRG DYADHQQWMG LSDSVRSCRL IPHSGSHRIR LYEREDYRGQ MIEFTEDCSC LQDRFRFNEI HSLNVLEGSW VLYELSNYRG RQYLLMPGDY RRYQDWGATN ARVGSLRRVI DFS
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications