missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GCC1 Protein

A DNA sequence encoding the Homo sapiens (Human) GCC1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7935-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031701

  • € 530.00 / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_078799.2
GCC1 Protein
Research Use Only
GCC1
Recombinant Protein
E. coli
FGMNFGGGPS KKDLLETIET QKKQLLQYQA RLKDVVRAYK SLLKEKEALE ASIKVLSVSH EADVGLAGVQ LPGLTFPDSV DDRCSTHSED STGTATSLDT AASLTSTKGE FGVEDDRPAR GPPPPKSEEA SWSESGVSSS SGDGPFAGGE VDKRLHQLKT QLATLTSSLA TVTQEKSRME ASYLADKKKM KQDLEDASNK AEEERARLEG ELKGLQEQIA ETKARLITQQ HDRAQEQSDH ALMLRELQKL LQEERTQRQD LELRLEETRE ALAGRAYAAE QMEGFELQTK QLTREVEELK SELQAIRDEK NQPDPRLQEL QEEAARLKSH FQAQLQQEMR KTALAEDQLR QQSQVEEQRV AALENQISEV SELLGTYEKA KQKDQLAIQK LKERILQLDL ENKTLALAAS SRSPLDSHGE ESSLDVNVLK DKMEKLKRLL QVAARKSQVT LDVEKLCDLE IMPSSEAADG EKATALYYQQ ELKQLKEEFE RYKMRAQVVL KSKNTKDGNL GKELEAAQEQ LAELKEKYIS LRLSCEELEH QHQQEADDWK QELARLQQLH RQELERCQLD FRDRTLKLEE ELHKQRDRAL AVLTEKDLEL EQLRSVALAS GLPGRRSPVG GGGPGDPADT SSSDSLTQAL QLAAANEPTF FLYAEQLARK EVEITSLRKQ KHRLEVEVHQ LQDRLLEEGE RHREEVAALQ SHIEKNIRDQ SREGANLEYL KNIIYRFLTL PDSLGRQQTL TAILTILHFS PEEKQVIMRL PTSASWWPSG KR
Greater than 90% as determined by SDS-PAGE.
79571
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers