missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDNF family receptor alpha-like Protein
A DNA sequence encoding the Homo sapiens (Human) GDNF family receptor alpha-like, was expressed in the hosts and tags indicated.
€ 245.00 - € 3405.00
Specifications
Accession Number | NP_997293.2 |
---|---|
Gene ID (Entrez) | 389400 |
Name | GDNF family receptor alpha-like Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16090591
|
enQuireBio™
QP7754-EC-100UG |
100 μg |
€ 770.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010601
|
enQuireBio™
QP7754-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16000601
|
enQuireBio™
QP7754-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020601
|
enQuireBio™
QP7754-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030601
|
enQuireBio™
QP7754-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040601
|
enQuireBio™
QP7754-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_997293.2 | |
GDNF family receptor alpha-like Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
389400 | |
Research Use Only | |
C6orf144 | |
Recombinant Protein | |
E. coli | |
SQTNNCTYLR EQCLRDANGC KHAWRVMEDA CNDSDPGDPC KMRNSSYCNL SIQYLVESNF QFKECLCTDD FYCTVNKLLG KKCINKSDNV KEDKFKWNLT TRSHHGFKGM WSCLEVAEAC VGDVVCNAQL ASYLKACSAN GNPCDLKQCQ AAIRFFYQNI PFNIAQMLAF CDCAQSDIPC QQSKEALHSK TCAVNMVPPP TCLSVIRSCQ NDELCRRHYR TFQSKCWQRV TRKCHEDENC ISTLSKQDLT CSGSDDCKAA YIDILGTVLQ VQCTCRTITQ SEESLCKIFQ HMLHRKSCFN YPTLSNVKGM ALYTRKHANK ITLTGFHSPF NGE | |
Greater than 90% as determined by SDS-PAGE. |