missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GDNF family receptor alpha-like Protein

A DNA sequence encoding the Homo sapiens (Human) GDNF family receptor alpha-like, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7754-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030601

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_997293.2
GDNF family receptor alpha-like Protein
Research Use Only
C6orf144
Recombinant Protein
E. coli
SQTNNCTYLR EQCLRDANGC KHAWRVMEDA CNDSDPGDPC KMRNSSYCNL SIQYLVESNF QFKECLCTDD FYCTVNKLLG KKCINKSDNV KEDKFKWNLT TRSHHGFKGM WSCLEVAEAC VGDVVCNAQL ASYLKACSAN GNPCDLKQCQ AAIRFFYQNI PFNIAQMLAF CDCAQSDIPC QQSKEALHSK TCAVNMVPPP TCLSVIRSCQ NDELCRRHYR TFQSKCWQRV TRKCHEDENC ISTLSKQDLT CSGSDDCKAA YIDILGTVLQ VQCTCRTITQ SEESLCKIFQ HMLHRKSCFN YPTLSNVKGM ALYTRKHANK ITLTGFHSPF NGE
Greater than 90% as determined by SDS-PAGE.
389400
500 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers