missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human General transcription factor IIF subunit 2 Protein
A DNA sequence encoding the Homo sapiens (Human) General transcription factor IIF subunit 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6132-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004119.1 | |
General transcription factor IIF subunit 2 Protein | |
Research Use Only | |
GTF2F2 | |
Recombinant Protein | |
E. coli | |
AERGELDLTG AKQNTGVWLV KVPKYLSQQW AKASGRGEVG KLRIAKTQGR TEVSFTLNED LANIHDIGGK PASVSAPREH PFVLQSVGGQ TLTVFTESSS DKLSLEGIVV QRAECRPAAS ENYMRLKRLQ IEESSKPVRL SQQLDKVVTT NYKPVANHQY NIEYERKKKE DGKRARADKQ HVLDMLFSAF EKHQYYNLKD LVDITKQPVV YLKEILKEIG VQNVKGIHKN TWELKPEYRH YQGEEKSD | |
Greater than 90% as determined by SDS-PAGE. |
2963 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |