missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GIGYF2 Protein
A DNA sequence encoding the Homo sapiens (Human) GIGYF2, was expressed in the hosts and tags indicated.
€ 202.00 - € 2155.00
Specifications
Accession Number | NP_001096616.1 |
---|---|
Gene ID (Entrez) | 26058 |
Name | GIGYF2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088761
|
enQuireBio™
QP9814-YE-100UG |
100 μg |
€ 645.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028771
|
enQuireBio™
QP9814-YE-500UG |
500 μg |
€ 1740.00
500µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098761
|
enQuireBio™
QP9814-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018771
|
enQuireBio™
QP9814-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038771
|
enQuireBio™
QP9814-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008771
|
enQuireBio™
QP9814-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001096616.1 | |
GIGYF2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
26058 | |
Research Use Only | |
GIGYF2 | |
Recombinant Protein | |
Yeast | |
ERLASKLQEH RAKGVSIPLM HEAMQKWYYK DPQGEIQGPF NNQEMAEWFQ AGYFTMSLLV KRACDESFQP LGDIMKMWGR VPFSPGPAPP PHMGELDQER LTRQQELTAL YQMQHLQYQQ FLIQQQYAQV LAQQQKAALS SQQQQQLALL LQQFQTLKMR ISDQNIIPSV TRSVSVPDTG SIWELQPTAS QPTVWEGGSV WDLPLDTTTP GPALEQLQQL EKAKAAKLEQ ERREAEMRAK REEEERKRQE ELRRQQEEIL RRQQEEERKR REEEELARRK QEEALRRQRE QEIALRRQRE EEERQQQEEA LRRLEERRRE EEERRKQEEL LRKQEEEAAK WAREEEEAQR RLEENRLRME EEAARLRHEE EERKRKELEV QRQKELMRQR QQQQEALRRL QQQQQQQQLA QMKLPSSSTW GQQSNTTACQ SQATLSLAEI QKLEEERERQ LREEQRRQQR ELMKALQQQQ QQQQQKLSGW GNVSKPSGTT KSLLEIQQEE ARQMQKQQQQ QQQHQQPNRA R | |
Greater than 90% as determined by SDS-PAGE. |