missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GITR/TNFRSF18 Fc Chimera Protein

Product Code. 18745516
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18745516

Brand: R&D Systems™ 689GR100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human (rh) GITR/TNFRSF18 Fc Chimera at 2 μg/mL (100 μL/well) can bind rhGITR Ligand with a linear range of 0.3-20 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y5U5
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8784
Molecular Weight (g/mol) M.W. (Observed): 50 kDa, reducing conditions; M.W. (theoretical): 41 kDa (monomer)
Name GITR/TNFRSF18 Fc Chimera
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived human GITR/TNFRSF18 protein Human GITR (Gln26-Glu161) (Thr45Ala) (N-terminus) Accession # Q9Y5U5 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol TNFRSF18
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein GITR/TNFRSF18
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.