missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glucokinase regulatory Protein

A DNA sequence encoding the Homo sapiens (Human) Glucokinase regulatory protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7596-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029640

  • € 2385.00 / 1mg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001477.2
Glucokinase regulatory Protein
Research Use Only
GCKR
Recombinant Protein
E. coli
MPGTKRFQHV IETPEPGKWE LSGYEAAVPI TEKSNPLTQD LDKADAENIV RLLGQCDAEI FQEEGQALST YQRLYSESIL TTMVQVAGKV QEVLKEPDGG LVVLSGGGTS GRMAFLMSVS FNQLMKGLGQ KPLYTYLIAG GDRSVVASRE GTEDSALHGI EELKKVAAGK KRVIVIGISV GLSAPFVAGQ MDCCMNNTAV FLPVLVGFNP VSMARNDPIE DWSSTFRQVA ERMQKMQEKQ KAFVLNPAIG PEGLSGSSRM KGGSATKILL ETLLLAAHKT VDQGIAASQR CLLEILRTFE RAHQVTYSQS PKIATLMKSV STSLEKKGHV YLVGWQTLGI IAIMDGVECI HTFGADFRDV RGFLIGDHSD MFNQKAELTN QGPQFTFSQE DFLTSILPSL TEIDTVVFIF TLDDNLTEVQ TIVEQVKEKT NHIQALAHST VGQTLPIPLK KLFPSIISIT WPLLFFEYEG NFIQKFQREL STKWVLNTVS TGAHVLLGKI LQNHMLDLRI SNSKLFWRAL AMLQRFSGQS KARCIESLLR AIHFPQPLSD DIRAAPISCH VQVAHEKEQV IPIALLSLLF RCSITEAQAH LAAAPSVCEA VRSALAGPGQ KRTADPLEIL EPDVQ
Greater than 80% as determined by SDS-PAGE.
2646
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers