missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glutamate receptor 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Glutamate receptor 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6120-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030950

  • € 2385.00 / 1mg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000819.3
Glutamate receptor 3 Protein
Research Use Only
GRIA3
Recombinant Protein
E. coli
SLLGHYKWEK FVYLYDTERG FSILQAIMEA AVQNNWQVTA RSVGNIKDVQ EFRRIIEEMD RRQEKRYLID CEVERINTIL EQVVILGKHS RGYHYMLANL
Greater than 90% as determined by SDS-PAGE.
2892
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers