missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glutathione S-transferase kappa 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Glutathione S-transferase kappa 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6125-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030990

Quantity Price
1 € 427.00 / 100µg
6 € 398.52 / 100µg
Estimated Shipment
22-10-2020
Add to basket


Specifications

Specifications

NP_001137151.1
Glutathione S-transferase kappa 1 Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
373156
Greater than 80% as determined by SDS-PAGE.
Research Use Only
GSTK1
Recombinant Protein
E. coli
GPLPRTVELF YDVLSPYSWL GFEILCRYQN IWNINLQLRP SLITGIMKDS GNKPPGLLPR KGLYMANDLK LLRHHLQIPI HFPKDFLSVM LEKGSLSAMR FLTAVNLEHP EMLEKASREL WMRVWSRNED ITEPQSILAA AEKAGMSAEQ AQGLLEKIAT PKVKNQLKET TEAACRYGAF GLPITVAHVD GQTHMLFGSD RMELLAHLLG EKWMGPIPPA VNARL
Videos
Documents