Learn More
enQuireBio™ Recombinant Human Glypican-6 Protein
Product Code. 16087741
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16087741
Brand: enQuireBio™ QP9160ye100ug
A DNA sequence encoding the Homo sapiens (Human) Glypican-6, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_005699.1
Gene ID (Entrez) 10082
Name Glypican-6 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol GPC6
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag His
Sequence DVKARSCGEV RQAYGAKGFS LADIPYQEIA GEHLRICPQE YTCCTTEMED KLSQQSKLEF ENLVEETSHF VRTTFVSRHK KFDEFFRELL ENAEKSLNDM FVRTYGMLYM QNSEVFQDLF TELKRYYTGG NVNLEEMLND FWARLLERMF QLINPQYHFS EDYLECVSKY TDQLKPFGDV PRKLKIQVTR AFIAARTFVQ GLTVGREVAN RVSKVSPTPG CIRALMKMLY CPYCRGLPTV RPCNNYCLNV MKGCLANQAD LDTEWNLFID AMLLVAERLE GPFNIESVMD PIDVKISEAI MNMQENSMQV SAKVFQGCGQ PKPAPALRSA RSAPENFNTR FRPYNPEERP TTAAGTSLDR LVTDIKEKLK LSKKVWSALP YTICKDESVT AGTSNEEECW NGHSKARYLP EIMNDGLTNQ INNPEVDVDI TRPDTFIRQQ IMALRVMTNK LKNAYNGNDV NFQDTSDESS GSGSGSGCMD DVCPTEFEFV TTEAPAVDPD RREVDS
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.