missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GMDS Protein

A cDNA sequence encoding the GMDS was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12001-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939058

  • € 120.00 / 5µg

Estimated Shipment: 02-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

2762
Greater than 95% as determined by SDS-PAGE.
Research Use Only
GMDS
Human
His
The GMDS solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl, 0.1mM PMSF and 30% glycerol.
GMDS Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAHAPARCPS ARGSGDGEMG KPRNVALITG ITGQDGSYLA EFLLEKGYEV HGIVRRSSSF NTGRIEHLYK NPQAHIEGNM KLHYGDLTDS TCLVKIINEV KPTEIYNLGA QSHVKISFDL AEYTADVDGV GTLRLLDAVK TCGLINSVKF YQASTSELYG KVQEIPQKET TPFYPRSPYG AAKLYAYWIV VNFREAYNLF AVNGILFNHE SPRRGANFVT RKISRSVAKI YLGQLECFSL GNLDAKRDWG HAKDYVEAMW LMLQNDEPED FVIATGEVHS VREFVEKSFL HIGKTIVWEG KNENEVGRCK ETGKVHVTVD LKYYRPTEVD FLQGDCTKAK QKLNWKPRVA FDELVREMVH ADVELMRTNP NA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers