missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNA15 Protein
A DNA sequence encoding the Homo sapiens (Human) GNA15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6100-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002059.3 | |
GNA15 Protein | |
Research Use Only | |
GNA15 | |
Recombinant Protein | |
E. coli | |
MARSLTWRCC PWCLTEDEKA AARVDQEINR ILLEQKKQDR GELKLLLLGP GESGKSTFIK QMRIIHGAGY SEEERKGFRP LVYQNIFVSM RAMIEAMERL QIPFSRPESK HHASLVMSQD PYKVTTFEKR YAAAMQWLWR DAGIRAYYER RREFHLLDSA VYYLSHLERI TEEGYVPTAQ DVLRSRMPTT GINEYCFSVQ KTNLRIVDVG GQKSERKKWI HCFENVIALI YLASLSEYDQ CLEENNQENR MKESLALFGT ILELPWFKST SVILFLNKTD ILEEKIPTSH LATYFPSFQG PKQDAEAAKR FILDMYTRMY TGCVDGPEGS KKGARSRRLF SHYTCATDTQ NIRKVFKDVR DSVLARYLDE INLL | |
Greater than 80% as determined by SDS-PAGE. |
2769 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-Trx | |
Tris-based buffer, 50% glycerol |