missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GNG12 Protein

A cDNA sequence encoding the GNG12 was constructed and used to recombinantly synthesize the protein.

€ 137.00 - € 4090.00

Specifications

Name GNG12 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15939108
View Documents Promotion Details
enQuireBio™
QP12017-5UG
5 μg
€ 137.00
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15929108
View Documents Promotion Details
enQuireBio™
QP12017-20UG
20 μg
€ 212.00
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15919108
View Documents Promotion Details
enQuireBio™
QP12017-1MG
1 mg
€ 4090.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

GNG12 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GNG12 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSSKTAS TNNIAQARRT VQQLRLEASI ERIKVSKASA DLMSYCEEHA RSDPLLIGIP TSENPFKDKK TC
Greater than 85.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications