missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNG12 Protein
A cDNA sequence encoding the GNG12 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12017-5ug
Additional Details : Weight : 0.01000kg
Specifications
GNG12 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSSKTAS TNNIAQARRT VQQLRLEASI ERIKVSKASA DLMSYCEEHA RSDPLLIGIP TSENPFKDKK TC | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GNG12 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |