missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GRB2 Protein

A cDNA sequence encoding the GRB2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12047-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939198

  • € 120.00 / 5µg

Estimated Shipment: 06-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

GRB2 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GRB2 protein solution contains 20mM Tris-HCl, pH 8 and 30% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MEAIAKYDFK ATADDELSFK RGDILKVLNE ECDQNWYKAE LNGKDGFIPK NYIEMKPHPW FFGKIPRAKA EEMLSKQRHD GAFLIRESES APGDFSLSVK FGNDVQHFKV LRDGAGKYFL WVVKFNSLNE LVDYHRSTSV SRNQQIFLRD IEQVPQQPTY VQALFDFDPQ EDGELGFRRG DFIHVMDNSD PNWWKGACHG QTGMFPRNYV TPVNRNV
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers