missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Growth / differentiation factor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Growth/differentiation factor 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9936-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057288.1 | |
Growth / differentiation factor 2 Protein | |
Research Use Only | |
GDF2 | |
Recombinant Protein | |
Yeast | |
HEEDTDGHVA AGSTLARRKR SAGAGSHCQK TSLRVNFEDI GWDSWIIAPK EYEAYECKGG CFFPLADDVT PTKHAIVQTL VHLKFPTKVG KACCVPTKLS PISVLYKDDM GVPTLKYHYE GMSVAECGCR | |
Greater than 90% as determined by SDS-PAGE. |
2658 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |