missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Growth / differentiation factor 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Growth/differentiation factor 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9936-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029041

  • € 1740.00 / 500µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_057288.1
Growth / differentiation factor 2 Protein
Research Use Only
GDF2
Recombinant Protein
Yeast
HEEDTDGHVA AGSTLARRKR SAGAGSHCQK TSLRVNFEDI GWDSWIIAPK EYEAYECKGG CFFPLADDVT PTKHAIVQTL VHLKFPTKVG KACCVPTKLS PISVLYKDDM GVPTLKYHYE GMSVAECGCR
Greater than 90% as determined by SDS-PAGE.
2658
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers