Learn More
DiscoverX Recombinant Human Growth Hormone
Product Code. 17312649
Encompass
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17312649 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17312649 Supplier DiscoverX Supplier No. 921338
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Recombinant Human Growth Hormone is an agonist for GHR. Provided as 100μg of dry powder alongside reconstitution buffer.
Human growth hormone (HGH, GH, somatotropin) is a 191-amino acid peptide pituitary hormone that stimulates growth, cell reproduction, and regeneration. An agonist for GHR, it also regulates body composition, fluids, and sugar and fat metabolism. It is a stress hormone that increases glucose and fatty acid levels and stimulates the production of IGF-1. Can be used medically to treat growth disorders and GH deficiency. Has been abused by competitors for its anabolic properties, and is banned by the IOC and NCAA. Provided as 100μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 100 μg
Product Type Control Ligands