missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GSK3B Protein

A DNA sequence encoding the Homo sapiens (Human) GSK3B, was expressed in the hosts and tags indicated.

€ 287.00 - € 3175.00

Specifications

Accession Number NP_001139628.1
Gene ID (Entrez) 2932
Name GSK3B Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16090960
View Documents Promotion Details
enQuireBio™
QP6121-YE-100UG
100 μg
€ 920.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16030970
View Documents Promotion Details
enQuireBio™
QP6121-YE-500UG
500 μg
€ 2650.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16000970
View Documents Promotion Details
enQuireBio™
QP6121-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16020970
View Documents Promotion Details
enQuireBio™
QP6121-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16040970
View Documents Promotion Details
enQuireBio™
QP6121-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16010970
View Documents Promotion Details
enQuireBio™
QP6121-YE-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001139628.1
GSK3B Protein
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
2932
Research Use Only
GSK3B
Recombinant Protein
Yeast
MSGRPRTTSF AESCKPVQQP SAFGSMKVSR DKDGSKVTTV VATPGQGPDR PQEVSYTDTK VIGNGSFGVV YQAKLCDSGE LVAIKKVLQD KRFKNRELQI MRKLDHCNIV RLRYFFYSSG EKKDEVYLNL VLDYVPETVY RVARHYSRAK QTLPVIYVKL YMYQLFRSLA YIHSFGICHR DIKPQNLLLD PDTAVLKLCD FGSAKQLVRG EPNVSYICSR YYRAPELIFG ATDYTSSIDV WSAGCVLAEL LLGQPIFPGD SGVDQLVEII KVLGTPTREQ IREMNPNYTE FKFPQIKAHP WTKVFRPRTP PEAIALCSRL LEYTPTARLT PLEACAHSFF DELRDPNVKL PNGRDTPALF NFTTQELSSN PPLATILIPP HARIQAAAST PTNATAASDA NTGDRGQTNN AASASASNST
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications