missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Guanylate cyclase activator 2B Protein

A DNA sequence encoding the Homo sapiens (Human) Guanylate cyclase activator 2B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7537-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029250

Quantity Price
1 € 2155.00 / 1mg
3 € 2011.05 / 1mg
Estimated Shipment
28-10-2020
Add to basket


Specifications

Specifications

NP_009033.1
Guanylate cyclase activator 2B Protein
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
2981
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Guca2b
Recombinant Protein
Yeast
VYIQYQGFRV QLESMKKLSD LEAQWAPSPR LQAQSLLPAV CHHPALPQDL QPVCASQEAS SIFKTLRTIA NDDCELCVNV ACTGCL
Videos
Documents