missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human H1F0 / Histone H1 Protein
A DNA sequence encoding the Homo sapiens (Human) H1F0 / Histone H1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6137-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005309.1 | |
H1F0 / Histone H1 Protein | |
Research Use Only | |
H1F0 | |
Recombinant Protein | |
E. coli | |
MTENSTSAP AAKPKRAKAS KKSTDHPKYS DMIVAAIQAE KNRAGSSRQS IQKYIKSHYK VGENADSQIK LSIKRLVTTG VLKQTKGVGA SGSFRLAKSD EPKKSVAFKK TKKEIKKVAT PKKASKPKKA ASKAPTKKPK ATPVKKAKKK LAATPKKAKK PKTVKAKPVK ASKPKKAKPV KPKAKSSAKR AGKKK | |
Greater than 80% as determined by SDS-PAGE. |
3005 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |