missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Hepsin Protein
Click to view available options
:
10ug; Carrier Free
Description
Measured by its ability to cleave tert-butoxycarbonyl-Gln-Arg-Arg-7-amino-4-methylcoumarin (Boc-QRR-AMC). The specific activity is >20,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P05981 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Acetate and NaCl. |
| Gene ID (Entrez) | 3249 |
| Molecular Weight (g/mol) | M.W. (Observed): 40-43 kDa, reducing conditions; M.W. (theoretical): 42 kDa |
| Name | Hepsin |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived human Hepsin protein Arg45-Leu417 (Asp161Glu & Arg162Lys), with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction