missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein A1 Protein

A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein A1, was expressed in the hosts and tags indicated.

€ 160.00 - € 2385.00

Specifications

Accession Number NP_002127.1
Gene ID (Entrez) 3178
Name Heterogeneous nuclear ribonucleoprotein A1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16091320
View Documents Promotion Details
enQuireBio™
QP6173-EC-100UG
100 μg
€ 530.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16011330
View Documents Promotion Details
enQuireBio™
QP6173-EC-1MG
1 mg
€ 2385.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16001330
View Documents Promotion Details
enQuireBio™
QP6173-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16021330
View Documents Promotion Details
enQuireBio™
QP6173-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16031330
View Documents Promotion Details
enQuireBio™
QP6173-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16041330
View Documents Promotion Details
enQuireBio™
QP6173-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_002127.1
Heterogeneous nuclear ribonucleoprotein A1 Protein
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
3178
Research Use Only
HNRNPA1
Recombinant Protein
E. coli
SKSESPKEPE QLRKLFIGGL SFETTDESLR SHFEQWGTLT DCVVMRDPNT KRSRGFGFVT YATVEEVDAA MNARPHKVDG RVVEPKRAVS REDSQRPGAH LTVKKIFVGG IKEDTEEHHL RDYFEQYGKI EVIEIMTDRG SGKKRGFAFV TFDDHDSVDK IVIQKYHTVN GHNCEVRKAL SKQEMASASS SQRGRSGSGN FGGGRGGGFG GNDNFGRGGN FSGRGGFGGS RGGGGYGGSG DGYNGFGNDG GYGGGGPGYS GGSRGYGSGG QGYGNQGSGY GGSGSYDSYN NGGGGGFGGG SGSNFGGGGS YNDFGNYNNQ SSNFGPMKGG NFGGRSSGPY GGGGQYFAKP RNQ
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications