missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Hexokinase-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Hexokinase-1, was expressed in the hosts and tags indicated.

€ 202.00 - € 2155.00

Specifications

Accession Number NP_000179.2
Gene ID (Entrez) 3098
Name Hexokinase-1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16091260
View Documents Promotion Details
enQuireBio™
QP6164-YE-100UG
100 μg
€ 645.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16031270
View Documents Promotion Details
enQuireBio™
QP6164-YE-500UG
500 μg
€ 1740.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16001270
View Documents Promotion Details
enQuireBio™
QP6164-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16021270
View Documents Promotion Details
enQuireBio™
QP6164-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16041270
View Documents Promotion Details
enQuireBio™
QP6164-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16011270
View Documents Promotion Details
enQuireBio™
QP6164-YE-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_000179.2
Hexokinase-1 Protein
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
3098
Research Use Only
HK1
Recombinant Protein
Yeast
ELKDDQVKKI DKYLYAMRLS DETLIDIMTR FRKEMKNGLS RDFNPTATVK MLPTFVRSIP DGSEKGDFIA LDLGGSSFRI LRVQVNHEKN QNVHMESEVY DTPENIVHGS GSQLFDHVAE CLGDFMEKRK IKDKKLPVGF TFSFPCQQSK IDEAILITWT KRFKASGVEG ADVVKLLNKA IKKRGDYDAN IVAVVNDTVG TMMTCGYDDQ HCEVGLIIGT GTNACYMEEL RHIDLVEGDE GRMCINTEWG AFGDDGSLED IRTEFDREID RGSLNPGKQL FEKMVSGMYL GELVRLILVK MAKEGLLFEG RITPELLTRG KFNTSDVSAI EKNKEGLHNA KEILTRLGVE PSDDDCVSVQ HVCTIVSFRS ANLVAATLGA ILNRLRDNKG TPRLRTTVGV DGSLYKTHPQ YSRRFHKTLR RLVPDSDVRF LLSESGSGKG AAMVTAVAYR LAEQHRQIEE TLA
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications