missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone deacetylase 6 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone deacetylase 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6153-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001308155.1 | |
Histone deacetylase 6 Protein | |
Research Use Only | |
HDAC6 | |
Recombinant Protein | |
E. coli | |
MTSTGQDSTT TRQRRSRQNP QSPPQDSSVT SKRNIKKGAV PRSIPNLAEV KKKGKMKKLG QAMEEDLIVG LQGMDLNLEA EALAGTGLVL DEQLNEFHCL WDDSFPEGPE RLHAIKEQLI QEGLLDRCVS FQARFAEKEE LMLVHSLEYI DLMETTQYMN EGELRVLADT YDSVYLHPNS YSCACLASGS VLRLVDAVLG AEIRNGMAII RPPGHHAQHS LMDGYCMFNH VAVAARYAQQ KHRIRRVLIV DWDVHHGQGT QFTFDQDPSV LYFSIHRYEQ GRFWPHLKAS NWSTTGFGQG QGYTINVPWN QVGMRDADYI AAFLHVLLPV ALEFQPQLVL VAAGFDALQG DPKGEMAATP AGFAQLTHLL MGLAGGKLIL SLEGGYNLRA LAEGVSASLH TLLGDPCPML ESPGAPCRSA QASVSCALEA LEPFWEVLVR STETVERDNM EEDNVEESEE EGPWEPPVLP ILTWPVLQSR TGLVYDQN | |
Greater than 85% as determined by SDS-PAGE. |
10013 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |