missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone deacetylase 6 Protein

A DNA sequence encoding the Homo sapiens (Human) Histone deacetylase 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6153-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051200

  • € 2385.00 / 1mg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001308155.1
Histone deacetylase 6 Protein
Research Use Only
HDAC6
Recombinant Protein
E. coli
MTSTGQDSTT TRQRRSRQNP QSPPQDSSVT SKRNIKKGAV PRSIPNLAEV KKKGKMKKLG QAMEEDLIVG LQGMDLNLEA EALAGTGLVL DEQLNEFHCL WDDSFPEGPE RLHAIKEQLI QEGLLDRCVS FQARFAEKEE LMLVHSLEYI DLMETTQYMN EGELRVLADT YDSVYLHPNS YSCACLASGS VLRLVDAVLG AEIRNGMAII RPPGHHAQHS LMDGYCMFNH VAVAARYAQQ KHRIRRVLIV DWDVHHGQGT QFTFDQDPSV LYFSIHRYEQ GRFWPHLKAS NWSTTGFGQG QGYTINVPWN QVGMRDADYI AAFLHVLLPV ALEFQPQLVL VAAGFDALQG DPKGEMAATP AGFAQLTHLL MGLAGGKLIL SLEGGYNLRA LAEGVSASLH TLLGDPCPML ESPGAPCRSA QASVSCALEA LEPFWEVLVR STETVERDNM EEDNVEESEE EGPWEPPVLP ILTWPVLQSR TGLVYDQN
Greater than 85% as determined by SDS-PAGE.
10013
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers