missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HNRNPC Protein
A cDNA sequence encoding the HNRNPC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12291-1ug
Additional Details : Weight : 0.01000kg
Specifications
HNRNPC Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MASNVTNKTD PRSMNSRVFI GNLNTLVVKK SDVEAIFSKY GKIVGCSVHK GFAFVQYVNE RNARAAVAGE DGRMIAGQVL DINLAAEPKV NRGKAGVKRS AAEMYGSSFD LDYDFQRDYY DRMYSYPARV PPPPPIARAV VPSKRQRVSG NTSRRGKSGF NSKSGQRGSS KSGKLKGDDL QAIKKELTQI KQKVDSLLEN LEKIEKEQSK QAVEMKNDKS EEEQSSSSVK KDETNVKMES EGGADDSAEE GDLLDDDDNE DRGDDQLELI KDDEKEAEEG EDDRDSANGE DDS | |
Greater than 85% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HNRNPC protein solution (0.25 mg/ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 5mM DTT and 30% glycerol. |