missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HNRNPC Protein

A cDNA sequence encoding the HNRNPC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12291-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939958

  • € 212.00 / 5µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HNRNPC Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MASNVTNKTD PRSMNSRVFI GNLNTLVVKK SDVEAIFSKY GKIVGCSVHK GFAFVQYVNE RNARAAVAGE DGRMIAGQVL DINLAAEPKV NRGKAGVKRS AAEMYGSSFD LDYDFQRDYY DRMYSYPARV PPPPPIARAV VPSKRQRVSG NTSRRGKSGF NSKSGQRGSS KSGKLKGDDL QAIKKELTQI KQKVDSLLEN LEKIEKEQSK QAVEMKNDKS EEEQSSSSVK KDETNVKMES EGGADDSAEE GDLLDDDDNE DRGDDQLELI KDDEKEAEEG EDDRDSANGE DDS
Greater than 85% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HNRNPC protein solution (0.25 mg/ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 5mM DTT and 30% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers