missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSP20 Protein
A cDNA sequence encoding the HSP20 was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 1905.00
Specifications
Name | HSP20 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940069
|
enQuireBio™
QP12326-2UG |
2 μg |
€ 137.00
2µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920069
|
enQuireBio™
QP12326-100UG |
100 μg |
€ 1905.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930069
|
enQuireBio™
QP12326-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
HSP20 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
HSP20 lyophilized from 20mM Tris-acetate, pH 7.6, 10mM NaCl, 0.1mM EDTA, 0.1mM PMSF and 15mM b-mercaptoethanol. |