missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSP20 Protein
Click to view available options
Quantity:
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
2µg
Specifications
Specifications
| Name | HSP20 Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | Untagged |
| Buffer | HSP20 lyophilized from 20mM Tris-acetate, pH 7.6, 10mM NaCl, 0.1mM EDTA, 0.1mM PMSF and 15mM b-mercaptoethanol. |
| Purity or Quality Grade | Greater than 95.0% as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction