missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSP20 Protein

A cDNA sequence encoding the HSP20 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12326-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940069

  • € 137.00 / 2µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HSP20 Protein
Research Use Only
Recombinant Protein
E. coli
HSP20 lyophilized from 20mM Tris-acetate, pH 7.6, 10mM NaCl, 0.1mM EDTA, 0.1mM PMSF and 15mM b-mercaptoethanol.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers