missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Hyaluronan-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Hyaluronan-binding protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7549-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001171131.1 | |
Hyaluronan-binding protein 1 | |
Research Use Only | |
HABP2 | |
Recombinant Protein | |
E. coli | |
WTPDQYDYSY EDYNQEENTS STLTHAENPD WYYTEDQADP CQPNPCEHGG DCLVHGSTFT CSCLAPFSGN KCQKVQNTCK DNPCGRGQCL ITQSPPYYRC VCKHPYTGPS CSQVVPVCRP NPCQNGATCS RHKRRSKFTC ACPDQFKGKF CEIGSDDCYV GDGYSYRGKM NRTVNQHACL YWNSHLLLQE NYNMFMEDAE THGIGEHNFC RNPDADEKPW CFIKVTNDKV KWEYCDVSAC SAQDVAYPEE SPTEPSTKLP GFDSCGKTEI AERKIKR | |
Greater than 90% as determined by SDS-PAGE. |
3026 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |