missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Hydroxymethylglutaryl-CoA lyase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Hydroxymethylglutaryl-CoA lyase, mitochondrial, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_000182.2 |
---|---|
Gene ID (Entrez) | 3155 |
Name | Hydroxymethylglutaryl-CoA lyase, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091290
|
enQuireBio™
QP6168-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011300
|
enQuireBio™
QP6168-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001300
|
enQuireBio™
QP6168-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021300
|
enQuireBio™
QP6168-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031300
|
enQuireBio™
QP6168-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041300
|
enQuireBio™
QP6168-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000182.2 | |
Hydroxymethylglutaryl-CoA lyase, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3155 | |
Research Use Only | |
HMGCL | |
Recombinant Protein | |
E. coli | |
TLPKRVKIVE VGPRDGLQNE KNIVSTPVKI KLIDMLSEAG LSVIETTSFV SPKWVPQMGD HTEVLKGIQK FPGINYPVLT PNLKGFEAAV AAGAKEVVIF GAASELFTKK NINCSIEESF QRFDAILKAA QSANISVRGY VSCALGCPYE GKISPAKVAE VTKKFYSMGC YEISLGDTIG VGTPGIMKDM LSAVMQEVPL AALAVHCHDT YGQALANTLM ALQMGVSVVD SSVAGLGGCP YAQGASGNLA TEDLVYMLEG LGIHTGVNLQ KLLEAGNFIC QALNRKTSSK VAQATCKL | |
Greater than 80% as determined by SDS-PAGE. |