missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ICAM2 Protein
A cDNA sequence encoding the ICAM2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12363-1mg
Additional Details : Weight : 0.01000kg
Specifications
ICAM2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSDEKVFE VHVRPKKLAV EPKGSLEVNC STTCNQPEVG GLETSLDKIL LDEQAQWKHY LVSNISHDTV LQCHFTCSGK QESMNSNVSV YQPPRQVILT LQPTLVAVGK SFTIECRVPT VEPLDSLTLF LFRGNETLHY ETFGKAAPAP QEATATFNST ADREDGHRNF SCLAVLDLMS RGGNIFHKHS APKMLEIYEP VSDSQ | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ICAM2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |