missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IER3 Protein
A cDNA sequence encoding the IER3 was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 1815.00
Specifications
Gene ID (Entrez) | 8870 |
---|---|
Name | IER3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | IER3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960199
|
enQuireBio™
QP12370-2UG |
2 μg |
€ 137.00
2µg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950199
|
enQuireBio™
QP12370-10UG |
10 μg |
€ 212.00
10µg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940199
|
enQuireBio™
QP12370-100UG |
100 μg |
€ 1815.00
100µg |
Estimated Shipment: Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
8870 | |
Research Use Only | |
IER3 | |
Human | |
His | |
IER3 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |
IER3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMCHSRSC HPTMTILQAP TPAPSTIPGP RRGSGPEIFT FDPLPEPAAA PAGRPSASRG HRKRSRRVLY PRVVRRQLPV EEPNP | |
Greater than 85.0% as determined by SDS-PAGE. |