missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IFN a 2b Protein

Product Code. 15964608
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15964608

Brand: enQuireBio™ QP10674EC100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the IFN a 2b was constructed and used to recombinantly synthesize the protein.

Specifications

Name IFN a 2b Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The specific activity was determined by the dose-dependent cytotoxicity of the human TF-1 cell line (human erythroleukemic indicator cell line) and is typically 0.02-0.08ng/ml.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag 20kD PEG
Sequence MCDLPQTHSL GSRRTLMLLA QMRRISLFSC LKDRHDFGFP QEEFGNQFQK AETIPVLHEM IQQIFNLFST KDSSAAWDET LLDKFYTELY QQLNDLEACV IQGVGVTETP LMKEDSILAV RKYFQRITLY LKEKKYSPCA WEVVRAEIMR SFSLSTNLQE SLRSKE
Buffer The IFN-a 2b was lyophilized from 1 mg/ml in 1xPBS.
Purity or Quality Grade Greater than 95% as obsereved by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.