missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human IFN-alpha 1 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Description
IFN-alpha 1 protein is fully biologically active when compared to standard. The specific activity determined by an anti-viral assay is no less than 1.0 x 10^8 IU/mg
Specifications
Specifications
| Accession Number | P01562 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS, pH 7.4, containing 3 % Mannitol, 0.05 % Tween-80. |
| Gene ID (Entrez) | 3439 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 19.5 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 0.1 EU/μg of IFN-alpha 1 AS determined by LAL method. |
| Gene Alias | 1, IFL;IFN;IFNA@;IFNA13;IFN-ALPHA;IFN-alphaD;Interferon alpha-1/13;leIF D |
| Gene Symbol | IFNA1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction